Snap Caps, Wheaton
Natural cap, 11 mm, PTFE/Silicone
Catalog # 89235-556
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 242776
Jump to:
Product Details & Documents
Eliminate the need for crimping and decapping tools.
- Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
Specifications
- Septum:PTFE/Silicone
- Désignation:Natural cap
- Taille:11 mm