Snap Caps, Wheaton
Natural cap, 11 mm, PTFE/Silicone
  89235-556
 :  242776
Snap Caps, Wheaton
  89235-556
Snap Caps, Wheaton
Snap Caps, Wheaton
Natural cap, 11 mm, PTFE/Silicone
1 263,50 $
Case of 1,000
Eliminate the need for crimping and decapping tools.
  • Manufactured from polyethylene
Fits the WHEATON E-Z vials® with snap ring top.
  • Septum:
    PTFE/Silicone
  • Désignation:
    Natural cap
  • Taille:
    11 mm