SmartCal™ Moisture Analyzer Test Substance | Avantor
SmartCal™ Moisture Analyzer Test Substance
Moisture analyzer test system starter pack (set of 12)
Catalog # CA11010-738
Supplier:   Mettler Toledo
Fournisseur #:  30005917
SmartCal™ Moisture Analyzer Test Substance
Catalog # CA11010-738
Supplier:  Mettler Toledo
SmartCal™ Moisture Analyzer Test Substance
SmartCal™ Moisture Analyzer Test Substance
Moisture analyzer test system starter pack (set of 12)
Price
$860.00
Log in pour la disponibilité
Each
Product Details & Documents
SmartCal™ is an innovative test substance that provides a fast way to verify the performance of a moisture analyzer.
  • Simply pour SmartCal™ into the sample pan and start
  • Test takes just 10 minutes
  • Easy to monitor and interpret instrument's performance over time
  • Store results directly in the instrument or manually enter into a free validated Excel® report sheet
  • Done just like a normal moisture measurement
  • Certified version available to satisfy regulatory requirements
  • SmartCal™ to get you up and running right away
SmartCal™ is a granular test substance. Its high sensitivity reliably detects adverse influences from ambient conditions and inaccurate heating or weighing devices.
Testing regularly with SmartCal™ ensures that moisture measurements are reliable at all times.
Documents
Spécifications
  • Désignation:
    Moisture analyzer test system starter pack (set of 12)