SmartCal™ Moisture Analyzer Test Substance
Moisture analyzer test system, certified version (set of 12)
Supplier: Mettler Toledo
Fournisseur #: 30005793
Jump to:
Product Details & Documents
SmartCal™ is an innovative test substance that provides a fast way to verify the performance of a moisture analyzer.
- Simply pour SmartCal™ into the sample pan and start
- Test takes just 10 minutes
- Easy to monitor and interpret instrument's performance over time
- Store results directly in the instrument or manually enter into a free validated Excel® report sheet
- Done just like a normal moisture measurement
- Certified version available to satisfy regulatory requirements
- SmartCal™ to get you up and running right away
SmartCal™ is a granular test substance. Its high sensitivity reliably detects adverse influences from ambient conditions and inaccurate heating or weighing devices.
Testing regularly with SmartCal™ ensures that moisture measurements are reliable at all times.
Documents
Certificates
Search by Lot NumberSpecifications
- Désignation:Moisture analyzer test system, certified version (set of 12)