EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences | Avantor
EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences
Improved Neubauer Double Cell 0.1 mm, Ruling allows full use
Catalog # 101765-030
Fournisseur #:  68052-16
EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences
Catalog # 101765-030
EMS Counting Cells for Haemocytometry, Electron Microscopy Sciences
Improved Neubauer Double Cell 0.1 mm, Ruling allows full use
Price
$500.78
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All complete with case and 1 pair of cover glasses.
The exceptions are the Howard Cell which doesn't include a cover glass and the Sedgewick Rafter which includes one cover glass.
Spécifications
  • Désignation:
    Improved Neubauer Double Cell 0.1 mm, Ruling allows full use