Lighted Forceps, Scissors, Electron Microscopy Sciences
Product Details & Documents
See your samples and your embeddings in a whole new light!
Increases the visibility of the specimen
Allows for accurate orientation of tissue and specimen
Minimizers floaters
Reduces embedding and prep time
These lighted forceps ease identification and location of otherwise undetectable tissue fragments, while reducing embedding time. Increases the visibility of specimen allowing for more accurate work. Made of German stainless steel, are submersible and light longevity approx. 20,000 hours.
Delivery information: Each kit includes: one instrument, power pack and battery charger.
Spécifications
- Désignation:Forceps
- Longueur:15 cm
- Tip Style:Smooth Fine Tip