StatMark Pen, Electron Microscopy Sciences | Avantor
StatMark Pen, Electron Microscopy Sciences
StatMark Pen
Catalog # 101764-994
Fournisseur #:  72109-12
StatMark Pen, Electron Microscopy Sciences
Catalog # 101764-994
StatMark Pen, Electron Microscopy Sciences
StatMark Pen, Electron Microscopy Sciences
StatMark Pen
Price
$147.15
Log in pour la disponibilité
Each Pack of 12
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes.
  • Permanent ink in an ultrafine tip
  • Quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes
  • Do not dry out easily
  • Last twice as long as many of the leading 'laboratory' pens
StatMark Pens are made of rigid nylon that allow for precise hand marking on glass slides as well as on most tissue embedding cassettes. This quick drying ink is highly resistant to aqueous based stains, alcohols, xylene and xylene substitutes. StatMark Pens do not dry out easily and last twice as long as many of the leading 'laboratory' pens.
Spécifications
  • Désignation:
    StatMark Pen