VWR® Buffer Solutions | Avantor
VWR® Buffer Solutions
Catalog # CA71006-816
VWR® Buffer Solutions
Catalog # CA71006-816
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BDH BUFFER REF STD PH10 BLUE 4L Catalog #BDH5076-4L
Le produit BDH5076-4L est similaire à CA71006-816 mais le fabricant et les spécifications peuvent varier.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.