VWR® Buffer Solutions
Catalog # CA71006-804
VWR® Buffer Solutions
Catalog # CA71006-804
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BDH BUFFER REF STD PH10 COLORLESS 4L Catalog #BDH5080-4L
Product BDH5080-4L is similar to CA71006-804 but manufacturer and specifications may vary.
BUFFER SOLN PH 1000 4L Catalog #CA1.09409.4000
Product CA1.09409.4000 is similar to CA71006-804 but manufacturer and specifications may vary.
BUFFER PH10 1GAL 1GL Catalog #RC16001
Product RC16001 is similar to CA71006-804 but manufacturer and specifications may vary.
Voir plus d'alternatives
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.