VWR® Buffer Solutions
VWR® Buffer Solutions
Catalog # CA71006-776
VWR® Buffer Solutions
Catalog # CA71006-776
Supplier:  
CAS Number:  
VWR® Buffer Solutions
VWR® Buffer Solutions
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BUFFER SOL PH 7.00 +0.01 2 4L Catalog #CAAA38711-K7
Product CAAA38711-K7 is similar to CA71006-776 but manufacturer and specifications may vary.
BUFFER PH 7 CALIBRATING 4L Catalog #RC1551-1
Product RC1551-1 is similar to CA71006-776 but manufacturer and specifications may vary.
BUFFER SOLN NBS REF PH7 4-L Catalog #CAJT5608-3
Product CAJT5608-3 is similar to CA71006-776 but manufacturer and specifications may vary.
Voir plus d'alternatives
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These buffer solutions are for calibrating pH meters
They are traceable to NIST standards.