WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
20 mm, Red
Catalog # 89220-260
 :  W242432-A
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Catalog # 89220-260
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
20 mm, Red
Price
$566.93
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 1,000
Product Details & Documents
Extended Controlled Dropper Tips are made of LDPE (Low-Density Polyethylene)
Extended Controlled Dropper Tips snap into the inside of the bottle neck and are available in bulk. Dropper tips can be covered with Wheaton dropper tip screw caps.
They dispense one drop at a time. The average drop size is 44µL (±6µL).
Spécifications
  • Taille du bouchon:
    20 mm
  • Color:
    Red