WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
20 mm, Pink
Catalog # 89220-256
Supplier: DWK Life Sciences (KIMBLE)
: W242431-A
WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Catalog # 89220-256
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
Extended Controlled Dropper Tips are made of LDPE (Low-Density Polyethylene)
Extended Controlled Dropper Tips snap into the inside of the bottle neck and are available in bulk. Dropper tips can be covered with Wheaton dropper tip screw caps.
They dispense one drop at a time. The average drop size is 44µL (±6µL).
Spécifications
- Taille du bouchon:20 mm
- Color:Pink