Caps for Mobile Phase Bottles, Microsolv
PP, Yellow, GL-45
Catalog # 89219-976
Supplier: MicroSolv Technology Corp
Supplier #: 58807-Y45
Jump to:
Product Details & Documents
These caps are designed for use on HPLC mobile phase reservoir and storage bottles with GL 45 thread.
- Made up of polypropylene
Specifications
- Color:Yellow
- Matière:PP
- Epaisseur:GL-45