Caps for Mobile Phase Bottles, Microsolv | Avantor
Caps for Mobile Phase Bottles, Microsolv
PP, Purple, GL-45
Catalog # 89219-974
Fournisseur #:  58807-P45
Caps for Mobile Phase Bottles, Microsolv
Catalog # 89219-974
Caps for Mobile Phase Bottles, Microsolv
Caps for Mobile Phase Bottles, Microsolv
PP, Purple, GL-45
Price
$98.68
Log in pour la disponibilité
Pack of 10
Product Details & Documents
These caps are designed for use on HPLC mobile phase reservoir and storage bottles with GL 45 thread.
  • Made up of polypropylene
Spécifications
  • Color:
    Purple
  • Matière:
    PP
  • Epaisseur:
    GL-45