Caps for Mobile Phase Bottles, Microsolv
PP, Blue, GL-45
  89219-970
 :  58807-B45
Caps for Mobile Phase Bottles, Microsolv
  89219-970
Caps for Mobile Phase Bottles, Microsolv
Caps for Mobile Phase Bottles, Microsolv
PP, Blue, GL-45
$98.68
Pack of 10
These caps are designed for use on HPLC mobile phase reservoir and storage bottles with GL 45 thread.
  • Made up of polypropylene
  • Color:
    Blue
  • Matière:
    PP
  • Epaisseur:
    GL-45