Caps for Mobile Phase Bottles, Microsolv
PP, Blue, GL-45
89219-970
: 58807-B45
These caps are designed for use on HPLC mobile phase reservoir and storage bottles with GL 45 thread.
- Made up of polypropylene
- Color:Blue
- Matière:PP
- Epaisseur:GL-45