Nunc® CryoColor Coder Inserts, Thermo Scientific
Blue
Supplier: Thermo Fisher Scientific
Supplier #: 354879
Nunc® CryoColor Coder Inserts, Thermo Scientific
Catalog # 24270-102
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
For comprehensive sample identification, fits inside CryoTube® vial cap.
- Made of polystyrene
- Available in 10 different colors
Ordering information: Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
Specifications
- Color:Blue