Ethylene Oxide Sterilization Indicator Labels
Ethylene Oxide Sterilization Indicator Labels
Plain, for Applicator Gun, 1000 Indicators per Roll
Catalog # 19872-030
Supplier:   Crosstex
Supplier #:  NMCPI-E02
Ethylene Oxide Sterilization Indicator Labels
Catalog # 19872-030
Supplier:  Crosstex
Ethylene Oxide Sterilization Indicator Labels
Ethylene Oxide Sterilization Indicator Labels
Plain, for Applicator Gun, 1000 Indicators per Roll
Price
$156.74
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Self-adhesive, nontoxic, chemical process indicators change from purple to green upon exposure to ethylene oxide.
Conform to AAMI ST60 and ISO 11140-1, Section 6 for Class 1 Process Indicators.
The color transition is sensitive to time, temperature, and presence of ethylene oxide. The exposure required for a color change is 600mg/L of ethylene oxide for a minimum of 30 minutes at 50–70% RH and 30°C (86°F). At longer exposure times or higher temperatures, the indicators exhibit a brighter shade of green. When exposed, imprinted indicators 19872-032 read “GREEN IS ETO EXPOSED.” Indicators have 24-month shelf life from date of manufacture when stored at room temperature.
Documents
Specifications
  • Désignation:
    Plain, for Applicator Gun, 1000 Indicators per Roll