Sample Cups for Abbott Analyzer, Globe Scientific | Avantor
Sample Cups for Abbott Analyzer, Globe Scientific
Sample Cup, 53 mm, 17.5 mm
Catalog # 60820-024
Supplier:   Globe Scientific
Fournisseur #:  5542
Sample Cups for Abbott Analyzer, Globe Scientific
Catalog # 60820-024
Supplier:  Globe Scientific
Sample Cups for Abbott Analyzer, Globe Scientific
Sample Cups for Abbott Analyzer, Globe Scientific
Sample Cup, 53 mm, 17.5 mm
Price
$146.15
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These sample cups and sample tubes are designed for use on Abbott® Axsym analyzers.
Spécifications
  • Ø ext.:
    17.5 mm
  • Désignation:
    Sample Cup
  • Hauteur:
    53 mm