VWR® Rapid Transfer Buffer, 10X
Rapid Transfer Buffer, 10X, 4 L
Catalog # 97064-314
   Safer Human and Environmental Health
Supplier:   VWR
 :  N789-4L
VWR® Rapid Transfer Buffer, 10X
Catalog # 97064-314
Supplier:  VWR
   Safer Human and Environmental Health
CAS Number:  
VWR® Rapid Transfer Buffer, 10X
VWR® Rapid Transfer Buffer, 10X
Rapid Transfer Buffer, 10X, 4 L
CAS Number:  
Price
$766.79
Log in pour la disponibilité
Each
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Rapid Transfer Buffer is a simple, single component system for quick and efficient transfer of proteins from SDS-PAGE gels to membranes for Western blotting applications
  • Fast, efficient transfer in 10–20 minutes
  • Utilizes standard transfer equipment
  • Non-hazardous formulation
  • Transfers to PVDF and nitrocellulose from multiple types of gels
Transfer is completed in 10 or 20 minutes using a standard semi-dry or wet transfer apparatus, respectively. Rapid Transfer Buffer is a methanol-free, non-hazardous formulation that is compatible with PVDF and nitrocellulose membranes. It is used for transfer of proteins from standard Laemmli gels, commercial pre-cast gels, and VWR® NEXT GEL™. The transfer efficiency is equivalent to that observed when using a Tris-Glycine-Methanol transfer buffer.
Spécifications
Spécifications
  • Désignation:
    Rapid Transfer Buffer, 10X
  • Taille:
    4 L
  • Cat. No.:
    97064-314
  • Environmentally Preferable:
    Safer Human and Environmental Health