VWR® RNase Removing Solution, Nucleaseliminator
VWR® RNase Removing Solution, Nucleaseliminator
NUCLEASELIMINATOR™, 100 mL
Catalog # 97064-070
Supplier:   VWR
Supplier #:  E891-100ML
VWR® RNase Removing Solution, Nucleaseliminator
Catalog # 97064-070
Supplier:  VWR
CAS Number:  
VWR® RNase Removing Solution, Nucleaseliminator
VWR® RNase Removing Solution, Nucleaseliminator
NUCLEASELIMINATOR™, 100 mL
CAS Number:  
Price
$63.25
Log in pour la disponibilité
Each (100ml)
Product Details & Documents
DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces.
  • Effectively removes and deactivates nuclease contamination even at high concentrations
  • Safe on all laboratory glass and plastic ware
  • Stable at room temperature for at least two years
Simply wipe, immerse or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips and electrophoresis units. NucleasEliminator™ is supplied as a ready-to-use liquid. A 100 ml bottle will treat up to 100 microcentrifuge tubes or 25 agarose electrophoresis casting trays.
Spécifications
Spécifications
  • Désignation:
    NUCLEASELIMINATOR™
  • Taille:
    100 mL
Spécifications