VWR® Tryptone
100 g
Catalog # 97063-386
Supplier:   VWR
 :  J859-100G
VWR® Tryptone
Catalog # 97063-386
Supplier:  VWR
CAS Number:  
VWR® Tryptone
VWR® Tryptone
100 g
CAS Number:  
Price
$88.84
Log in pour la disponibilité
Each (100g) Case of 12 (100g)
Product Details & Documents
High-purity pancreatic protein digests of casein used for media supplements.
  • High-purity
  • Pancreatic protein digests
  • Used for media supplements
High-purity pancreatic protein digests of casein used for media supplements.
Spécifications
Spécifications
  • Taille:
    100 g
Spécifications