VWR® MOPS Buffer Solution 10X, Biotechnology Grade | Avantor
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
100 mL
Catalog # 97062-852
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
Catalog # 97062-852
Supplier:  
CAS Number:  
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
100 mL
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Optimized for RNA Agarose Electrophoresis
Ideal for Use with RNA Formaldehyde Gels.
Spécifications
Spécifications
  • Taille:
    100 mL