VWR® SSC Buffer
SSC BUFFER, 20X LIQUID CONCENTRATE, 4 L, Hybridization buffer.
Catalog # 97062-390
Supplier:   VWR
 :  0804-4L
VWR® SSC Buffer
Catalog # 97062-390
Supplier:  VWR
CAS Number:  
VWR® SSC Buffer
VWR® SSC Buffer
SSC BUFFER, 20X LIQUID CONCENTRATE, 4 L, Hybridization buffer.
CAS Number:  
Price
$284.81
Log in pour la disponibilité
Each (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SSC Buffer is a hybridization buffer for Northern and Southern blots
VWR® offers SSC in various convenient forms: a 20X liquid concentrate, a tablet that prepares 100 ml of a 1X solution, and a Ready-Pack™ that prepares 1 L of 20X concentrate.
Spécifications
Spécifications
  • Désignation:
    SSC BUFFER, 20X LIQUID CONCENTRATE
  • Taille:
    4 L
  • Utilisation:
    Hybridization buffer.