Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
VWR® Malt Extract

VWR® Malt Extract

Catalog # 97063-428
VWR® Malt Extract
Catalog # 97063-428
Supplier:  
CAS Number:  
VWR® Malt Extract
VWR® Malt Extract
CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

VWR® offers high purity extracts used for media supplements.

  • Used for the preparation of media for the detection of yeasts and molds. Hazards, N/A. Bacteriological Grade
Specifications

Specifications

  • Cat. No.:
    97063-428
  • Total nitrogen:
    2.0%
  • Storage conditions:
    Cold
  • Ash content:
    5%
  • Solubility:
    6%, Water, Pass
  • pH:
    6% Water, 4.5 to 5.7 (at 25 °C)
  • Coliforms:
    10/g
  • Loss on drying:
    5.0%

Specifications