Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
NIST Traceable pH Buffers with Temperature Table

NIST Traceable pH Buffers with Temperature Table

Cubetainer®, 10 L, 10.00
Catalog # 97007-806
Supplier:   Mettler Toledo
Supplier #:  63056126
NIST Traceable pH Buffers with Temperature Table
Catalog # 97007-806
Supplier:  Mettler Toledo
CAS Number:  
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table

Cubetainer®, 10 L, 10.00

CAS Number:  
Price
$162.00
Log in for availability
Each (10l)
Minimum order quantity is 1
Product Details & Documents

NIST-traceable buffer for pH electrode calibration and verification.

  • Temperature table on bottle

Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.

Documents

Specifications

Specifications

  • Type de conditionnement:
    Cubetainer®
  • Taille:
    10 L
  • pH:
    10.00

Specifications