Reference Standards for Food Analysis, Restek
Reference Standards for Food Analysis, Restek
Methyl Heneicosanoate, C21:0, 100 mg
Catalog # RK35061
Reference Standards for Food Analysis, Restek
Catalog # RK35061
Supplier:  
CAS Number:  
Reference Standards for Food Analysis, Restek
Reference Standards for Food Analysis, Restek
Methyl Heneicosanoate, C21:0, 100 mg
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Neat fatty acid methyl esters are used to prepare specific mixtures.
  • Each compound is packaged under a nitrogen blanket to ensure product stability.
Specifications
Specifications
  • Désignation:
    Methyl Heneicosanoate, C21:0, 100 mg