Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Rtx®-50 Phase Columns, Restek

Rtx®-50 Phase Columns, Restek

Capillaire, 0.50 µm, 0.53 mm, 15 m
Catalog # RK10537
Rtx®-50 Phase Columns, Restek
Catalog # RK10537
Supplier:  
Rtx®-50 Phase Columns, Restek
Rtx®-50 Phase Columns, Restek

Capillaire, 0.50 µm, 0.53 mm, 15 m

Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
GSBP-50+MS 15M 0.53MM 0.50UM Catalog #10499-710

Product 10499-710 is similar to RK10537 but manufacturer and specifications may vary.

Product Details & Documents

For analysis of pesticides, herbicides, phthalate esters, cholesterols, triglycerides, and rosin acids.

  • Low bleed
  • Longer column lifetime
  • Polymer is fully characterized to ensure long-term reproducibility

Can be used as a confirmational column for many environmental applications including EPA methods 604 (phenols) and 8060 (phthalate esters) and for the analysis of EPA method 608 (pesticides).

Fused silica columns are coated with crossbonded 50% methyl/50% phenyl polysiloxane stationary phase. The high thermal stability of these columns allows dual column analysis with common phases such as Rtx®-1 and Rtx®-5 for simultaneous run conditions and high column bake-out temperatures to remove contamination. Restek's Crossbond® procedure produces columns with lower bleed, longer lifetime, and column rejuvenation through solvent washing. No residual catalyst remains in the polymer which could cause degradation and increase bleed. The polymer is carefully fractionated to remove low molecular weight fragments and provide tight mono-modal distribution which results in extremely low bleed.

Spécifications
  • Type de colonne:
    Capillaire
  • Épaisseur du film:
    0.50 µm
  • Ø int.:
    0.53 mm
  • Longueur:
    15 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.