Multiple Affinity Removal Column HSA/IgG, Agilent Technologies
Affinity Removal System Column, Human Albumin/IgG, 4.60 mm, 50 mm
Catalog # 97066-658
Supplier: Agilent Technologies
Fournisseur #: 5188-8826
Multiple Affinity Removal Column HSA/IgG, Agilent Technologies
Catalog # 97066-658
Supplier: Agilent Technologies
Jump to:
Product Details & Documents
The multiple affinity removal LC column – human albumin/IgG is designed to chromatographically remove the two most interfering high-abundant proteins (albumin and IgG) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
- Serum injection volume is 100 µl per run for the 4.6 mm id column
- Optimize sample recovery by utilizing rapid, single injection protocols and optimized buffers
- Column is compatible with most FPLC or HPLC pumps or instruments
Removal of albumin and IgG improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.
Spécifications
- Ø int.:4.60 mm
- Désignation:Affinity Removal System Column, Human Albumin/IgG
- Longueur:50 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



