Rapidblock™ Blocking Solution, 10X, AMRESCO
Product Details & Documents
Solution reduces blocking time to 5 minutes for Western and dot blotting procedures on PVDF and nitrocellulose membranes.
- Only 5 Minutes for PVDF or Nitrocellulose Blocking
- Reduce Background and Cross-Reactivity
- Enhance Chemiluminescent Signals
- Protein-Free Formulation
This protein-free formulation minimizes cross-reactivity and non-specific antibody binding, generating blots with low background and enhanced signal-to-noise ratio. Results meet or exceed those obtained with buffers containing dried milk or BSA, which both require 1 hour of blocking time.
Documents
Specifications
Specifications
- Taille:15 mL