VWR® SSC Buffer
VWR® SSC Buffer
SSC BUFFER, 20X READY-PACK™, 2 pack, Each Ready-Pack™ prepares 1L of 20X concentrate.
Catalog # 97062-382
VWR® SSC Buffer
Catalog # 97062-382
Supplier:  
CAS Number:  
VWR® SSC Buffer
VWR® SSC Buffer
SSC BUFFER, 20X READY-PACK™, 2 pack, Each Ready-Pack™ prepares 1L of 20X concentrate.
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
BUFFER SSC HYBRIDIZATION 20X 4L Catalog #97062-390
Product 97062-390 is similar to 97062-382 but manufacturer and specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SSC Buffer is a hybridization buffer for Northern and Southern blots
VWR® offers SSC in various convenient forms: a 20X liquid concentrate, a tablet that prepares 100 ml of a 1X solution, and a Ready-Pack™ that prepares 1 L of 20X concentrate.
Spécifications
Spécifications
  • Désignation:
    SSC BUFFER, 20X READY-PACK™
  • Taille:
    2 pack
  • Utilisation:
    Each Ready-Pack™ prepares 1L of 20X concentrate.