Multi-Purpose Sampling Cups, Globe Scientific | Avantor
Multi-Purpose Sampling Cups, Globe Scientific
0.25 ml (0.008 oz.), Polystyrene, 14 × 16 mm, Clear
Catalog # 60820-054
Supplier:   Globe Scientific
Fournisseur #:  110711
Multi-Purpose Sampling Cups, Globe Scientific
Catalog # 60820-054
Supplier:  Globe Scientific
Multi-Purpose Sampling Cups, Globe Scientific
Multi-Purpose Sampling Cups, Globe Scientific
0.25 ml (0.008 oz.), Polystyrene, 14 × 16 mm, Clear
Price
$81.38
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Transparent, polystyrene cups are durable and chemical-resistant
  • Polystyrene (PS)
  • Can be used on most common brands of instruments
They feature a wide base design to prevent tipping or spilling and a conical bottom for use in microsampling. A variety of cup sizes permits use with most autoanalyzers. Cups are designed to accept snap caps (not included).
Spécifications
  • Capacité:
    0.25 ml (0.008 oz.)
  • Color:
    Clear
  • Matière:
    Polystyrene
  • Diameter ×H:
    14 × 16 mm