Analyzer-Specific Sample Cups, Globe Scientific | Avantor
Analyzer-Specific Sample Cups, Globe Scientific
Blue
Catalog # 60819-996
Supplier:   Globe Scientific
Fournisseur #:  6500B
Analyzer-Specific Sample Cups, Globe Scientific
Catalog # 60819-996
Supplier:  Globe Scientific
Analyzer-Specific Sample Cups, Globe Scientific
Analyzer-Specific Sample Cups, Globe Scientific
Blue
Price
$111.91
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Polyethylene sample cups are for use with Cobas Mira® analyzers.
Spécifications
  • Color:
    Blue