Simultaneous Test Sets, Drager Safety
Simultaneous test set II, for inorganic fumes (sulfur dioxide, chlorine, hydrogen sulfide, hydrogen phosphide, carbonyl chloride), 40 s
  13453-126
 :   Draeger
 :  8101736
Simultaneous Test Sets, Drager Safety
  13453-126
 :  Draeger
Simultaneous Test Sets, Drager Safety
Simultaneous Test Sets, Drager Safety
Simultaneous test set II, for inorganic fumes (sulfur dioxide, chlorine, hydrogen sulfide, hydrogen phosphide, carbonyl chloride), 40 s
$164.37
Each
Risks and potential dangers at a glance: The SimultanTest Sets allow the measurement of five different gases at the same time.
In the SimultanTest Set, five Dräger-Tubes are arranged next to each other in a stable rubber sleeve. The air to be monitored is simultaneously drawn in through the tubes by a Dräger-Tube pump. Quick determinations are made about the presence of dangerous substances at TLV levels.
The device delivers fast and reliable results for different combinations of organic and inorganic gases.
  • Désignation:
    Simultaneous test set II, for inorganic fumes (sulfur dioxide, chlorine, hydrogen sulfide, hydrogen phosphide, carbonyl chloride)
  • Response Time:
    40 s
Accessories
  •  
    Description
  • CA13450-858
    Extension Hose
    Each - $1,769.37
  •  
    Description
  • CA34330-044
    Adapter for Simultaneous test sets, for connection to a bellows pump
    Each - $595.27