Electrode Solutions
Electrode Solutions
FRISCOLYT-B® electrolyte, 6×250 ml, Bottle
Catalog # 11278-356
Supplier:   Mettler Toledo
Supplier #:  51350084
Electrode Solutions
Catalog # 11278-356
Supplier:  Mettler Toledo
CAS Number:  
Electrode Solutions
Electrode Solutions
FRISCOLYT-B® electrolyte, 6×250 ml, Bottle
CAS Number:  
Price
$208.00
Log in for availability
Each (250ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made of highest quality reagents to provide optimum performance in all types of applications.
Documents
Specifications
Specifications
  • Type de conditionnement:
    Bottle
  • Désignation:
    FRISCOLYT-B® electrolyte
  • Taille:
    6×250 ml