Reference Standards for Food Analysis, Restek
Methyl Nonanoate, C9:0, 100 mg
Catalog # RK35040
Reference Standards for Food Analysis, Restek
Catalog # RK35040
Supplier:  
CAS Number:  
Reference Standards for Food Analysis, Restek
Reference Standards for Food Analysis, Restek
Methyl Nonanoate, C9:0, 100 mg
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Neat fatty acid methyl esters are used to prepare specific mixtures.
  • Each compound is packaged under a nitrogen blanket to ensure product stability.
Spécifications
Spécifications
  • Désignation:
    Methyl Nonanoate, C9:0, 100 mg