High-Temperature String, Restek | Avantor
High-Temperature String, Restek
High-Temperature String
Catalog # RK20109
Supplier:   Restek
Fournisseur #:  20109
High-Temperature String, Restek
Catalog # RK20109
Supplier:  Restek
High-Temperature String, Restek
High-Temperature String, Restek
High-Temperature String
Price
$73.45
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Use high-temperature string to restring capillary columns, attach column connectors to column cages, or hold the column in the GC oven.
Capable of withstanding temperatures to 400 °C.
Spécifications
  • Désignation:
    High-Temperature String