Microsurgical Knife, Electron Microscopy Sciences
Micro Surgical Knife 15°
  101413-486
 :  72047-15
Microsurgical Knife, Electron Microscopy Sciences
  101413-486
Microsurgical Knife, Electron Microscopy Sciences
Microsurgical Knife, Electron Microscopy Sciences
Micro Surgical Knife 15°
$186.14
Each Pack of 6
Made from tool steel wire. Sharpened to microsurgical quality.
The knife-edges are available in three different configurations: 45, 30 and 15 degrees angle. The handle is plastic and they come sterile and in a blister pack individually.
 : 6 blister packs to a box.
  • Désignation:
    Micro Surgical Knife 15°