General Purpose Transfer Pipette, Electron Microscopy Sciences
Product Details & Documents
These pipettes are good for blood banking, reagent handling, viscous liquid sampling and transferring, etc. The non-sterile pipettes are bulk packed and the sterile are individually packed.
Spécifications
- Ø×L:6.6×15.4 cm
- Sterility:Non sterile
- Volume:4.0 ml
- Bulb Draw:1.9 ml