Xylene Substitute, Electron Microscopy Sciences
This EMS Xylene Substitute solution is specially designed for use in processing and staining histological/cytological specimens. It is essentially odorless with low toxicity, less irritating to the skin than xylene- and d-limonene-based clearants. It can be substituted for xylene and can be used with almost all tissue processors and automated slide stainers.
- Minimal tissue shrinkage
- Dries slowly with no residue
- Less toxic than xylene
- Soluble with alcohol, paraffin embedding media, and mounting media
- Biodegradable
Xylene substitute for histology and cytology is a safer alternative to xylene. Can be used as a solvent and clearing agent, as well as for dissolving paraffin waxes, glues and adhesives. It can be used in all procedures that require xylene.
- Type de conditionnement:Bottle
- Désignation:Xylene substitute
- Taille:1 gal.