HANKS's Balanced Salts, Electron Microscopy Sciences
HANKS's Balanced Salts, Electron Microscopy Sciences
500 ml
Catalog # 100502-386
Minority or Woman-Owned Business Enterprise
Supplier #:  11768-05
HANKS's Balanced Salts, Electron Microscopy Sciences
Catalog # 100502-386
Supplier:  
Minority or Woman-Owned Business Enterprise
CAS Number:  
HANKS's Balanced Salts, Electron Microscopy Sciences
500 ml
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All of our Hanks’ Balanced Salt Solutions contain: Potassium chloride, potassium phosphate monobasic, sodium chloride, sodium phosphate dibasic, and D-glucose. Some formulations may contain additional components. Use the table below as a quick reference for identifying the one that best suits your application.
All solutions are prepared in 1X strength.
Spécifications
Spécifications
  • Taille:
    500 ml
  • Cat. No.:
    100502-386
  • Size:
    500 ml