Count Up/Count Down Timer, Electron Microscopy Sciences
Timer, count up count down
  100502-098
Count Up/Count Down Timer, Electron Microscopy Sciences
  100502-098
Count Up/Count Down Timer, Electron Microscopy Sciences
Count Up/Count Down Timer, Electron Microscopy Sciences
Timer, count up count down
$79.37
Each
Count up or down for 100 hours in intervals of 1 second.
  • Alarm clock mode
  • Its memory allows for the repeat of a cycle just completed
In the count down mode its unique feature tells how much time has elapsed after the alarm sounds.
Dimensions: 2³/₄×2³/₄×¹/₂"
Weight: 1.8 oz.
 : With N.I.S.T. traceable certificate of compliance.
 : Comes complete with a built in stand, pocket clip, button cell battery, and magnet.
  • Désignation:
    Timer, count up count down