Syringe Filtration System, Electron Microscopy Sciences
Product Details & Documents
Syrings filters with high burst strength for assured performance. Manufactured to ISO 9002 and GMP standard and 100% integrity tested.
- For tissue culture and HPLC applications
- Membrane choices include:
- Regenerated cellulose (RC)
- Polyethersulfone (PES)
- Surfactant-free cellulose acetate (SFCA)
- Nylon (NY)
- 'PTFE' fluourocarbon resin
Syringe filter sizes include 4, 15, 25, or 26 mm diameter to handle volumes ranging from 0.5 to 100 ml. The 50 mm diameter 'PTFE' syringe filter are ideal for protecting the vacuum line and pump. These disposable filter units are ideal for tissue culture media, buffer solution, stain solution, biological fluids and most other aqueous solutions and reagents that are used daily in the EM lab.
Pore size and membrane type are printed on every syringe and the filters are certified non-cytotoxic and non-pyrogenic.
Packaging: Sterile syringe filters are individually blister-wrapped in cases of 50. Non sterile syringe filters are packaged in cases of 50. The 50 mm 'PTFE' syringe filter is supplied in a case of 12.
Spécifications
- Ø:25 mm
- Matériau du boîtier:PP
- Taille de pore:0.45 µm
- Désignation:Syringe Filter "PTFE"