All Plastic Syringes, Electron Microscopy Sciences
All plastic syringes are the choice for any situation needing an inert, hypoallergenic syringe. Because of their purity, they are indicated for IVF, embryo transfer, chromatography and many laboratory procedures.
- Latex free and contain no rubber, no silicone oil or styrene
- Manufactured only from laboratory grade polypropylene and polyethylene
- Unique plastic syringes have a positive safety stop to prevent accidental spills
: These syringes are individually sterile strip packed.
- Type:Eccentric
- Color:Clear
- Désignation:Syringe 10 cc