Vibration Isolation Platform, Electron Microscopy Sciences | Avantor
Vibration Isolation Platform, Electron Microscopy Sciences
Platform, 110 - 190 lbs., 18×24"
Catalog # 100498-968
Fournisseur #:  67125-18
Vibration Isolation Platform, Electron Microscopy Sciences
Catalog # 100498-968
Vibration Isolation Platform, Electron Microscopy Sciences
Vibration Isolation Platform, Electron Microscopy Sciences
Platform, 110 - 190 lbs., 18×24"
Price
$9,735.88
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
VIP Series 300 isolation technology is superior to air technology. First, these vibration isolation platforms are passive, mechanical and maintenance free. There are no air hoses or supporting hardware required. Second, Vistek bearings isolate effectively for the horizontal and vertical vibration components.
  • Custom shapes, sizes and colors are available on request
  • Bearings are nickel-plated alloy steel with clear anodized aluminum
  • Plate is black anodized aluminum
  • The payload ranges from 35/8 to 31/8 and 25/8
The superior performance of the Vistek bearings in all six degrees of freedom makes this isolation platform the best choice for eliminating unwanted vibrations. Virtually any type of scientific instrument may be positioned on the Series 300 Vibration Isolation Platform. Standard sizes are 12 x 18", 18 x 24", and 24 x 30", for general use.
Units such as microscopes, hardness testers, measuring devices, digital imaging systems, AFMs, and Analytical balances are all applications for a bench top or tabletop platform. Transmissibility Curve for the VIP Series 300 1/6th Hertz Vibration Isolation Bearing. The natural frequency of horizontal platforms is 0.3 Hz, and vertical platforms vary with the payload.
Spécifications
  • Désignation:
    Platform
  • W×D:
    18×24"
  • Weighing Capacity:
    110 - 190 lbs.