Petroff-Hausser Chambers, Electron Microscopy Sciences
Product Details & Documents
New improved Neubauer ruling pattern with 1/400 sq. mm of ruling. Designed for the counting of bacteria, sperm, blood platelets, and the content of vaccines.
Cell is 0.02 mm deep with 1.5 mm thick glass slide allows the use off dark field microscopy.
Spécifications
- Désignation:Petroff Hausser chamber, no grid with case (3900sp)