Fine Diamond Scriber, Electron Microscopy Sciences
Bent, fine point diamond scriber, 60° point angle, 0.5 mm
Catalog # 100497-836
 :  62107-BT
Fine Diamond Scriber, Electron Microscopy Sciences
Catalog # 100497-836
Fine Diamond Scriber, Electron Microscopy Sciences
Fine Diamond Scriber, Electron Microscopy Sciences
Bent, fine point diamond scriber, 60° point angle, 0.5 mm
Price
$106.59
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This diamond scriber is the ultimate scribing instrument offering the most precision for the most delicate applications.
  • Fine scribing under the microscope - for silicon wafers and glass coverslips
  • Precision scribing and repairing - for thin film circuits and microcircuits, and more
It is available in straight or bent configurations.The integrated 6.35 mm anodized hexagonal (non-rolling) aluminum handle has a positive finger grip in the form of grooves for easy control and nonslippage. Each diamond scriber is packaged in a protective plastic tube.
Spécifications
  • Désignation:
    Bent, fine point diamond scriber, 60° point angle
  • Diameter:
    0.5 mm