Diamond Lapping Film, Electron Microscopy Sciences
Product Details & Documents
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis. Comes in 8" and 12" discs with plain back (P/B) or pressure sensitive adhesive back (PSA).
Spécifications
- Granulométrie:9 µm
- Taille:12"
- Backing:Plain back