Diamond Lapping Film, Electron Microscopy Sciences
Diamond Lapping Film, Electron Microscopy Sciences
0.1 µm, Plain back, 8"
  100497-196
Supplier #:  50350-01
Diamond Lapping Film, Electron Microscopy Sciences
  100497-196
Diamond Lapping Film, Electron Microscopy Sciences
Diamond Lapping Film, Electron Microscopy Sciences
0.1 µm, Plain back, 8"
96,40 $
Each
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis. Comes in 8" and 12" discs with plain back (P/B) or pressure sensitive adhesive back (PSA).
  • Granulométrie:
    0.1 µm
  • Taille:
    8"
  • Backing:
    Plain back