ESD Safe Tweezers, Electron Microscopy Sciences | Avantor
ESD Safe Tweezers, Electron Microscopy Sciences
Style 3C SA/Dr, SA steel, 110 mm, Very sharp and superior finish
Catalog # 100494-844
Fournisseur #:  78393-3C
ESD Safe Tweezers, Electron Microscopy Sciences
Catalog # 100494-844
ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
Style 3C SA/Dr, SA steel, 110 mm, Very sharp and superior finish
Price
108,66 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
Spécifications
  • Désignation:
    Style 3C SA/Dr
  • Matière:
    SA steel
  • Tip Style:
    Very sharp and superior finish
  • Overall Length:
    110 mm