ESD Safe Tweezers, Electron Microscopy Sciences | Avantor
ESD Safe Tweezers, Electron Microscopy Sciences
Style 00 SA/Dr, SA steel, 120 mm, Flat edges and strong tips
Catalog # 100494-838
Fournisseur #:  78390-00
ESD Safe Tweezers, Electron Microscopy Sciences
Catalog # 100494-838
ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
Style 00 SA/Dr, SA steel, 120 mm, Flat edges and strong tips
Price
105,69 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
Spécifications
  • Désignation:
    Style 00 SA/Dr
  • Matière:
    SA steel
  • Tip Style:
    Flat edges and strong tips
  • Overall Length:
    120 mm